missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl Methacrylate 98.0+%, TCI America™
$294.52 - $1174.35
Chemical Identifiers
| CAS | 38862-25-8 |
|---|---|
| Molecular Formula | C8H9NO4 |
| Molecular Weight (g/mol) | 183.16 |
| MDL Number | MFCD15072193 |
| InChI Key | ACGJEMXWUYWELU-UHFFFAOYSA-N |
| Synonym | Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide |
| PubChem CID | 10130244 |
| IUPAC Name | 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate |
| SMILES | CC(=C)C(=O)ON1C(=O)CCC1=O |
Chemical Identifiers
| 38862-25-8 | |
| 183.16 | |
| ACGJEMXWUYWELU-UHFFFAOYSA-N | |
| 10130244 | |
| CC(=C)C(=O)ON1C(=O)CCC1=O |
| C8H9NO4 | |
| MFCD15072193 | |
| Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide | |
| 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate |
Specifications
| 38862-25-8 | |
| White | |
| MFCD15072193 | |
| Methacrylic Acid N-Succinimidyl Ester, N-Methacryloyloxysuccinimide | |
| CC(=C)C(=O)ON1C(=O)CCC1=O | |
| 183.16 | |
| 183.16 | |
| Crystalline Powder |
| 104°C | |
| C8H9NO4 | |
| 5 g | |
| ACGJEMXWUYWELU-UHFFFAOYSA-N | |
| 2,5-dioxopyrrolidin-1-yl 2-methylprop-2-enoate | |
| 10130244 | |
| ≥98.0% (GC) | |
| N-Succinimidyl Methacrylate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator