missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl Ferrocenecarboxylate 98.0+%, TCI America™
$227.41 - $689.70
Chemical Identifiers
| CAS | 115223-09-1 |
|---|---|
| Molecular Formula | C15H15FeNO4 |
| Molecular Weight (g/mol) | 329.133 |
| MDL Number | MFCD03427488 |
| InChI Key | CNVNCOWBDQLYOU-UHFFFAOYSA-N |
| Synonym | Ferrocenecarboxylic Acid N-Succinimidyl Ester |
| PubChem CID | 14226536 |
| IUPAC Name | cyclopenta-1,3-diene;(2,5-dioxopyrrolidin-1-yl) cyclopenta-1,3-diene-1-carboxylate;iron |
| SMILES | C1CC(=O)N(C1=O)OC(=O)C2=CC=CC2.C1C=CC=C1.[Fe] |
Chemical Identifiers
| 115223-09-1 | |
| 329.133 | |
| CNVNCOWBDQLYOU-UHFFFAOYSA-N | |
| 14226536 | |
| C1CC(=O)N(C1=O)OC(=O)C2=CC=CC2.C1C=CC=C1.[Fe] |
| C15H15FeNO4 | |
| MFCD03427488 | |
| Ferrocenecarboxylic Acid N-Succinimidyl Ester | |
| cyclopenta-1,3-diene;(2,5-dioxopyrrolidin-1-yl) cyclopenta-1,3-diene-1-carboxylate;iron |
Specifications
| 115223-09-1 | |
| MFCD03427488 | |
| Ferrocenecarboxylic Acid N-Succinimidyl Ester | |
| C1CC(=O)N(C1=O)OC(=O)C2=CC=CC2.C1C=CC=C1.[Fe] | |
| 329.133 | |
| 327.12 | |
| Crystalline Powder |
| C15H15FeNO4 | |
| 200 mg | |
| CNVNCOWBDQLYOU-UHFFFAOYSA-N | |
| cyclopenta-1,3-diene;(2,5-dioxopyrrolidin-1-yl) cyclopenta-1,3-diene-1-carboxylate;iron | |
| 14226536 | |
| ≥98.0% (HPLC) | |
| N-Succinimidyl Ferrocenecarboxylate |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator