missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl Ferrocenecarboxylate 98.0+%, TCI America™

Catalog No. S0820200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0820200MG 200 mg
S08201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0820200MG Supplier TCI America Supplier No. S0820200MG

Chemical Identifiers

CAS 115223-09-1
Molecular Formula C15H15FeNO4
Molecular Weight (g/mol) 329.133
MDL Number MFCD03427488
InChI Key CNVNCOWBDQLYOU-UHFFFAOYSA-N
Synonym Ferrocenecarboxylic Acid N-Succinimidyl Ester
PubChem CID 14226536
IUPAC Name cyclopenta-1,3-diene;(2,5-dioxopyrrolidin-1-yl) cyclopenta-1,3-diene-1-carboxylate;iron
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC=CC2.C1C=CC=C1.[Fe]

Specifications

Quantity 200 mg
Formula Weight 327.12
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Ferrocenecarboxylate
TSCA No
Recommended Storage Refrigerator