missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl Acrylate 98.0+%, TCI America™

Catalog No. S081425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S081425G 25 g
S08145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S081425G Supplier TCI America Supplier No. S081425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 38862-24-7
Molecular Formula C7H7NO4
Molecular Weight (g/mol) 169.14
MDL Number MFCD00078261
InChI Key YXMISKNUHHOXFT-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl acrylate, n-acryloxysuccinimide, n-succinimidyl acrylate, acrylic acid n-hydroxysuccinimide ester, 1-acryloyloxy-2,5-pyrrolidinedione, n-acryloyloxysuccinimide, 2,5-pyrrolidinedione, 1-1-oxo-2-propenyl oxy, 2,5-dioxopyrrolidin-1-yl prop-2-enoate, poly n-acryloxysuccinimide
PubChem CID 181508
IUPAC Name 2,5-dioxopyrrolidin-1-yl prop-2-enoate
SMILES C=CC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 25 g
Formula Weight 169.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl Acrylate
TSCA No
Recommended Storage Freezer