accessibility menu, dialog, popup

UserName

H2COONOO

CAS RN 38862-24-7

IUPAC Name: 2,5-dioxopyrrolidin-1-yl prop-2-enoate
Synonyms: 2,5-dioxopyrrolidin-1-yl prop-2-enoate
Molecular Weight (g/mol): 169.14
Molecular Formula: C7H7NO4
InChi Key: YXMISKNUHHOXFT-UHFFFAOYSA-N
SMILES: C=CC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)