missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate 96.0+%, TCI America™

$296.38 - $891.02

Chemical Identifiers

CAS 134471-24-2
Molecular Formula C14H9NO7
Molecular Weight (g/mol) 303.226
MDL Number MFCD00037568
InChI Key KFEBWCYYRFZMTJ-UHFFFAOYSA-N
Synonym 7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester
PubChem CID 5706554
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 7-hydroxy-2-oxochromene-3-carboxylate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C(C=C3)O)OC2=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S0866200MG
View Documents
TCI America
S0866200MG
200 mg
Each for $296.38
Only null left
 
S08661G
View Documents
TCI America
S08661G
1 g
Each for $891.02
Only null left
 
Description

Description

Specifications

Chemical Identifiers

134471-24-2
303.226
KFEBWCYYRFZMTJ-UHFFFAOYSA-N
5706554
C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C(C=C3)O)OC2=O
C14H9NO7
MFCD00037568
7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester
(2,5-dioxopyrrolidin-1-yl) 7-hydroxy-2-oxochromene-3-carboxylate

Specifications

134471-24-2
C14H9NO7
200 mg
KFEBWCYYRFZMTJ-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 7-hydroxy-2-oxochromene-3-carboxylate
5706554
≥96.0% (HPLC,T)
N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate
Yellow
MFCD00037568
7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester
C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C(C=C3)O)OC2=O
303.226
303.23
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Freezer

SDS
missing translation for 'documents'

missing translation for 'documents'