accessibility menu, dialog, popup

UserName

ONOOOOHOO

CAS RN 134471-24-2

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 7-hydroxy-2-oxo-2H-chromene-3-carboxylate
Synonyms: 2,5-dioxopyrrolidin-1-yl 7-hydroxy-2-oxochromene-3-carboxylate
Molecular Weight (g/mol): 303.23
Molecular Formula: C14H9NO7
InChi Key: KFEBWCYYRFZMTJ-UHFFFAOYSA-N
SMILES: OC1=CC=C2C=C(C(=O)ON3C(=O)CCC3=O)C(=O)OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results