missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate 96.0+%, TCI America™

Catalog No. S0866200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0866200MG 200 mg
S08661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0866200MG Supplier TCI America Supplier No. S0866200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 134471-24-2
Molecular Formula C14H9NO7
Molecular Weight (g/mol) 303.226
MDL Number MFCD00037568
InChI Key KFEBWCYYRFZMTJ-UHFFFAOYSA-N
Synonym 7-Hydroxycoumarin-3-carboxylic Acid N-Succinimidyl Ester, N-Succinimidyl Umbelliferone-3-carboxylate, Umbelliferone-3-carboxylic Acid N-Succinimidyl Ester
PubChem CID 5706554
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 7-hydroxy-2-oxochromene-3-carboxylate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC3=C(C=C(C=C3)O)OC2=O

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 303.23
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 7-Hydroxycoumarin-3-carboxylate
TSCA No
Recommended Storage Freezer