missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Succinimidyl 7-Hydroxy-4-methylcoumarin-3-acetate 90.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Quantity | 25 mg |
| Formula Weight | 331.28 |
| Percent Purity | ≥90.0% (HPLC,N) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Succinimidyl 7-Hydroxy-4-methylcoumarin-3-acetate |
Safety and Handling
| TSCA | No |
| Recommended Storage | Freezer (-20 °C) |