accessibility menu, dialog, popup

UserName

CAS RN 96735-88-5

CH3OONOOOOOH

CAS RN 96735-88-5

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-(7-hydroxy-4-methyl-2-oxo-2H-chromen-3-yl)acetate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2-(7-hydroxy-4-methyl-2-oxochromen-3-yl)acetate
Molecular Weight (g/mol): 331.28
Molecular Formula: C16H13NO7
InChi Key: VODLZKHEKKGCBG-UHFFFAOYSA-N
SMILES: CC1=C(CC(=O)ON2C(=O)CCC2=O)C(=O)OC2=CC(O)=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results