missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 6-Biotinamidohexanoate 97.0+%, TCI America™

Catalog No. S0490100MG
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0490100MG 100 mg
S049020MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0490100MG Supplier TCI America Supplier No. S0490100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72040-63-2
Molecular Formula C20H30N4O6S
Molecular Weight (g/mol) 454.542
MDL Number MFCD00065502
InChI Key UVGHPGOONBRLCX-NJSLBKSFSA-N
Synonym 6-Biotinamidohexanoic Acid N-Succinimidyl Ester, N-[5-(N-Succinimidyloxycarbonyl)pentyl]biotinamide, N-Biotinyl-6-aminohexanoic Acid Succinimidyl Ester
PubChem CID 83874
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]hexanoate
SMILES C1CC(=O)N(C1=O)OC(=O)CCCCCNC(=O)CCCCC2C3C(CS2)NC(=O)N3

Specifications

Melting Point 170°C
Color White
Quantity 100 mg
Formula Weight 454.54
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 6-Biotinamidohexanoate
TSCA No
Recommended Storage Freezer