accessibility menu, dialog, popup

UserName

CAS RN 72040-63-2

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 6-{5-[(3aS,4S,6aR)-2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl]pentanamido}hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 6-{5-[(3aS,4S,6aR)-2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl]pentanamido}hexanoate
Molecular Weight (g/mol): 454.54
Molecular Formula: C20H30N4O6S
InChi Key: UVGHPGOONBRLCX-NJSLBKSFSA-N
SMILES: O=C(CCCC[C@@H]1SC[C@@H]2NC(=O)N[C@H]12)NCCCCCC(=O)ON1C(=O)CCC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results