missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]hexanoate 98.0+%, TCI America™

Catalog No. s088125mg
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S088125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S088125MG Supplier TCI America Supplier No. S088125MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 125559-00-4
Molecular Formula C22H29N3O7
Molecular Weight (g/mol) 447.49
MDL Number MFCD01863459
InChI Key IHVODYOQUSEYJJ-UHFFFAOYSA-N
Synonym N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]caproate, LC-SMCC
PubChem CID 3564731
IUPAC Name 2,5-dioxopyrrolidin-1-yl 6-({4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexyl}formamido)hexanoate
SMILES O=C(CCCCCNC(=O)C1CCC(CN2C(=O)C=CC2=O)CC1)ON1C(=O)CCC1=O

Specifications

Melting Point 182°C
Color Yellow
Quantity 25 mg
Formula Weight 447.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 6-[[4-(N-Maleimidomethyl)cyclohexyl]carboxamido]hexanoate
TSCA No
Recommended Storage Freezer