accessibility menu, dialog, popup

UserName

CAS RN 125559-00-4

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 6-({4-[(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)methyl]cyclohexyl}formamido)hexanoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 6-({4-[(2,5-dioxopyrrol-1-yl)methyl]cyclohexyl}formamido)hexanoate
Molecular Weight (g/mol): 447.49
Molecular Formula: C22H29N3O7
InChi Key: IHVODYOQUSEYJJ-UHFFFAOYSA-N
SMILES: O=C(CCCCCNC(=O)C1CCC(CN2C(=O)C=CC2=O)CC1)ON1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results