missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 3-Maleimidobenzoate 98.0+%, TCI America™

Catalog No. S0398100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0398100MG 100 mg
S03981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0398100MG Supplier TCI America Supplier No. S0398100MG

Chemical Identifiers

CAS 58626-38-3
Molecular Formula C15H10N2O6
Molecular Weight (g/mol) 314.253
MDL Number MFCD00005514
InChI Key LLXVXPPXELIDGQ-UHFFFAOYSA-N
Synonym 3-maleimidobenzoic acid n-hydroxysuccinimide ester, n-succinimidyl 3-maleimidobenzoate, 3-maleimidobenzoyl n-hydroxysuccinimide, 1-3-2,5-dioxopyrrolidinyl oxy carbonyl phenyl-1h-pyrrole-2,5-dione, m-maleimidobenzoyl-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl benzoate, 3-n-maleimidobenzoic acid n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzoate, ambotzpeg2090
PubChem CID 93861
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)benzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC(=CC=C2)N3C(=O)C=CC3=O

Specifications

Melting Point 181°C
Color White-Yellow
Quantity 100 mg
Formula Weight 314.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Succinimidyl 3-Maleimidobenzoate [Cross-linking Reagent]
TSCA No
Recommended Storage Refrigerator