missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl 3-maleimidobenzoate, 95%

Catalog No. AAJ6141003
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6141003 1 g
AAJ61410MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6141003 Supplier Thermo Scientific Chemicals Supplier No. J6141003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A heterobifunctional coupling reagent useful for forming enzyme immunoconjugates | CAS: 58626-38-3 | C15H10N2O6 | 314.253 g/mol

N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Succinimidyl 3-maleimidobenzoate is useful for forming enzyme immunoconjugates. It has a 9.9 Angstrom spacer arm. Heterobifunctional crosslinking reagent reactive toward primary amine and sulfhydryl. Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates

Solubility
Soluble in DMF or ethyl acetate.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 58626-38-3
Molecular Formula C15H10N2O6
Molecular Weight (g/mol) 314.253
MDL Number MFCD00005514
InChI Key LLXVXPPXELIDGQ-UHFFFAOYSA-N
Synonym 3-maleimidobenzoic acid n-hydroxysuccinimide ester, n-succinimidyl 3-maleimidobenzoate, 3-maleimidobenzoyl n-hydroxysuccinimide, 1-3-2,5-dioxopyrrolidinyl oxy carbonyl phenyl-1h-pyrrole-2,5-dione, m-maleimidobenzoyl-n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxopyrrol-1-yl benzoate, 3-n-maleimidobenzoic acid n-succinimidyl ester, 2,5-dioxopyrrolidin-1-yl 3-2,5-dioxo-2,5-dihydro-1h-pyrrol-1-yl benzoate, ambotzpeg2090
PubChem CID 93861
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3-(2,5-dioxopyrrol-1-yl)benzoate
SMILES C1CC(=O)N(C1=O)OC(=O)C2=CC(=CC=C2)N3C(=O)C=CC3=O

Specifications

Melting Point 175°C to 177°C
Color White
Quantity 1 g
Beilstein 1505254
Sensitivity Light and moisture sensitive
Solubility Information Soluble in DMF or ethyl acetate.
Formula Weight 314.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Succinimidyl 3-maleimidobenzoate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 261-368-8
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only