missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Salicylideneaniline 98.0+%, TCI America™
$42.87 - $160.30
Chemical Identifiers
| CAS | 779-84-0 |
|---|---|
| Molecular Formula | C13H11NO |
| Molecular Weight (g/mol) | 197.237 |
| MDL Number | MFCD00020066 |
| InChI Key | GVHXCXKLYZOBKS-UHFFFAOYSA-N |
| Synonym | n-salicylideneaniline, phenol, 2-e-phenylimino methyl, n-salicylalaniline, 2-hydroxybenzalaniline, o-hydroxy benzylidene aniline, acmc-20m6ov, acmc-1bijs, n-o-hydroxybenzylidene aniline, 2-e-phenyliminomethyl phenol, phenol,2-phenylimino methyl |
| PubChem CID | 6740751 |
| IUPAC Name | 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one |
| SMILES | C1=CC=C(C=C1)NC=C2C=CC=CC2=O |
Chemical Identifiers
| 779-84-0 | |
| 197.237 | |
| GVHXCXKLYZOBKS-UHFFFAOYSA-N | |
| 6740751 | |
| C1=CC=C(C=C1)NC=C2C=CC=CC2=O |
| C13H11NO | |
| MFCD00020066 | |
| n-salicylideneaniline, phenol, 2-e-phenylimino methyl, n-salicylalaniline, 2-hydroxybenzalaniline, o-hydroxy benzylidene aniline, acmc-20m6ov, acmc-1bijs, n-o-hydroxybenzylidene aniline, 2-e-phenyliminomethyl phenol, phenol,2-phenylimino methyl | |
| 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one |
Specifications
| 779-84-0 | |
| 151°C | |
| MFCD00020066 | |
| n-salicylideneaniline, phenol, 2-e-phenylimino methyl, n-salicylalaniline, 2-hydroxybenzalaniline, o-hydroxy benzylidene aniline, acmc-20m6ov, acmc-1bijs, n-o-hydroxybenzylidene aniline, 2-e-phenyliminomethyl phenol, phenol,2-phenylimino methyl | |
| C1=CC=C(C=C1)NC=C2C=CC=CC2=O | |
| 197.237 | |
| 197.24 | |
| Crystalline Powder |
| 51°C | |
| C13H11NO | |
| 5 g | |
| GVHXCXKLYZOBKS-UHFFFAOYSA-N | |
| 6-(anilinomethylidene)cyclohexa-2,4-dien-1-one | |
| 6740751 | |
| ≥98.0% (GC,T) | |
| N-Salicylideneaniline |
Safety and Handling
TSCA : No