missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine 98.0+%, TCI America™
$42.87 - $124.89
Chemical Identifiers
| CAS | 82717-96-2 |
|---|---|
| Molecular Formula | C15H21NO4 |
| Molecular Weight (g/mol) | 279.34 |
| MDL Number | MFCD00209976 |
| InChI Key | CEIWXEQZZZHLDM-AAEUAGOBSA-N |
| Synonym | s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine |
| PubChem CID | 5702571 |
| IUPAC Name | (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid |
| SMILES | CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O |
Chemical Identifiers
| 82717-96-2 | |
| 279.34 | |
| CEIWXEQZZZHLDM-AAEUAGOBSA-N | |
| 5702571 | |
| CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O |
| C15H21NO4 | |
| MFCD00209976 | |
| s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine | |
| (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid |
Specifications
| 82717-96-2 | |
| White | |
| MFCD00209976 | |
| s-2-s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, n-s-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, unii-10sz4ij77b, n-1 s-ethoxycarbonyl-3-phenylpropyl-l-alanine, n-s-+-1-ethoxycarbonyl-3-phenylpropyl-l-alanine, 2s-2-2s-1-ethoxy-1-oxo-4-phenylbutan-2-yl amino propanoic acid, s,s-n-ethoxy carbonyl-3-phenylpropyl alanine, n-s-1-carboxyethyl-l-homophenylalanine ethyl ester, s,s-n-1-ethoxycarbonyl-3-phenylpropyl alanine | |
| CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(O)=O | |
| 279.34 | |
| 279.34 | |
| Crystalline Powder |
| 150°C | |
| C15H21NO4 | |
| 5 g | |
| CEIWXEQZZZHLDM-AAEUAGOBSA-N | |
| (2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoic acid | |
| 5702571 | |
| ≥98.0% (HPLC,T) | |
| N-[(S)-1-Ethoxycarbonyl-3-phenylpropyl]-L-alanine |
Safety and Handling
EINECSNumber : (3)-3931
TSCA : No