missing translation for 'onlineSavingsMsg'
Learn More

n-Propyltriethoxysilane, 97%

CAS: 2550-02-9 | C9H22O3Si | 206.36 g/mol

$233.01 - $865.00

Chemical Identifiers

CAS 2550-02-9
MDL Number MFCD00026756
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2099722
View Documents
Thermo Scientific Chemicals
B2099722
100 g
Each for $233.01
Only null left
 
AAB2099736
View Documents
Thermo Scientific Chemicals
B2099736
500 g
Each for $865.00
Only null left
 
Description

Description

It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, aromatic and aliphatic hydrocarbons.

Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications

Chemical Identifiers

2550-02-9
MFCD00026756

Specifications

179°C to 180°C
100 g
MFCD00026756
1748158
propyltriethoxysilane, triethoxy propyl silane, n-propyltriethoxysilane, silane, triethoxypropyl, unii-f17otx9fg5, f17otx9fg5, dynasylan pteo, triethoxypropyl-silan, triethoxypropyl-silane, triethoxy-n-propylsilane
NBXZNTLFQLUFES-UHFFFAOYSA-N
triethoxy(propyl)silane
75691
97%
Moisture Sensitive
n-Propyltriethoxysilane
2550-02-9
C9H22O3Si
UN1993
57°C (134°F)
Not miscible or difficult to mix in water. Soluble in alcohol,aromatic and aliphatic hydrocarbons.
CCC[Si](OCC)(OCC)OCC
206.36
206.36
1.3965
0.8916
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 219-842-7

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only