Learn More
n-Propyltriethoxysilane, 97%
CAS: 2550-02-9 | C9H22O3Si | 206.36 g/mol
Supplier: Thermo Scientific Chemicals B2099736
Description
It is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important material in sol-gel production, it provides a hydrophobic surface treatment for inorganic powders or filter materials and it is one of the catalyst components for polyolefin production with Ziegler-Natta catalyst. It is mainly used as construction waterproofing agent/protective agent, inorganic filler surface treatment agent, and pigment dispersant.
Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, aromatic and aliphatic hydrocarbons.
Notes
Moisture sensitive. Store away from air and oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Keep ignition sources away.
Specifications
| 179°C to 180°C | |
| 2550-02-9 | |
| C9H20O3Si | |
| UN1993 | |
| 57°C (134°F) | |
| Not miscible or difficult to mix in water. Soluble in alcohol,aromatic and aliphatic hydrocarbons. | |
| CCO[Si](CC=C)(OCC)OCC | |
| 204.34 | |
| 206.36 | |
| 1.3965 | |
| 0.8916 |
| n-Propyltriethoxysilane | |
| 500 g | |
| MFCD00026756 | |
| 1748158 | |
| allyltriethoxysilane, silane, triethoxy-2-propenyl, triethoxyallylsilane, silane, allyltriethoxy, allytriethoxysilane, triethoxy prop-2-en-1-yl silane, silane, triethoxy-2-propen-1-yl, allyl triethoxysilane, acmc-2097bb | |
| UMFJXASDGBJDEB-UHFFFAOYSA-N | |
| triethoxy(prop-2-enyl)silane | |
| 75692 | |
| 97% | |
| Moisture Sensitive |
Chemical Identifiers
| 2550-02-9 | |
| MFCD00026756 |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 219-842-7
TSCA : Yes
RUO – Research Use Only