missing translation for 'onlineSavingsMsg'
Learn More

N-Propylaniline 98.0+%, TCI America™

Catalog No. P062625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P062625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P062625ML Supplier TCI America Supplier No. P062625ML
Only null left

Chemical Identifiers

CAS 622-80-0
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
MDL Number MFCD00048678
InChI Key CDZOGLJOFWFVOZ-UHFFFAOYSA-N
PubChem CID 12153
IUPAC Name N-propylaniline
SMILES CCCNC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 220°C
Quantity 25 mL
Formula Weight 135.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Propylaniline
EINECSNumber (3)-0118&(3)-0206
TSCA Yes