accessibility menu, dialog, popup

UserName

H3CNH

CAS RN 622-80-0

IUPAC Name: N-propylaniline
Synonyms: N-propylaniline
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: CDZOGLJOFWFVOZ-UHFFFAOYSA-N
SMILES: CCCNC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results