missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl formate, 97%

Catalog No. AAA18090AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA18090AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA18090AP Supplier Thermo Scientific Chemicals Supplier No. A18090AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 110-74-7 | C4H8O2 | 88.106 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-74-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00003295
InChI Key KFNNIILCVOLYIR-UHFFFAOYSA-N
Synonym formic acid, propyl ester, n-propyl formate, propyl methanoate, formiate de propyle, formic acid propyl ester, propylester kyseliny mravenci, unii-wo1arv6gtw, formiate de propyle french, fema no. 2943, wo1arv6gtw
PubChem CID 8073
IUPAC Name propyl formate
SMILES CCCOC=O

Specifications

Melting Point -93°C
Density 0.904
Boiling Point 80°C to 81°C
Flash Point −4°C (25°F)
Refractive Index 1.375
Quantity 500 mL
UN Number UN1281
Beilstein 1739272
Merck Index 14,7858
Formula Weight 88.11
Percent Purity 97%
Chemical Name or Material n-Propyl formate
Show More Show Less
Hazard Category H225-H319-H335-H336
Hazard Statement GHS H Statement
H225-H319-H335-H336
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYL FORMATES
EINECSNumber 203-798-0
RTECSNumber LR0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only