missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl decanoate, 98%

Catalog No. p-7028043
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2515918 50 g
AAB2515930 250 g
AAB251590B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2515918 Supplier Thermo Scientific Chemicals Supplier No. B2515918

CAS: 30673-60-0 | C13H26O2 | 214.349 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 30673-60-0
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.349
MDL Number MFCD00056329
InChI Key OVFMRFMJVFDSAA-UHFFFAOYSA-N
Synonym n-propyl decanoate, decanoic acid, propyl ester, propyl caprate, capric acid propyl ester, unii-s0he9grs1w, decanoic acid, propylester, s0he9grs1w, propyl caprate natural, propyl caprate decanoate, n-propyldecanoate
PubChem CID 121739
ChEBI CHEBI:86973
IUPAC Name propyl decanoate
SMILES CCCCCCCCCC(=O)OCCC

Specifications

Density 0.86
Boiling Point 254°C
Flash Point 115°C (239°F)
Quantity 50 g
Formula Weight 214.35
Percent Purity 98%
Chemical Name or Material n-Propyl decanoate
EINECSNumber 250-283-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only