missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Propyl Alcohol, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5%, Honeywell Burdick & Jackson
$2785.70 - $2785.70
Chemical Identifiers
| CAS | 71-23-8 |
|---|---|
| Molecular Formula | C3H8O |
| Molecular Weight (g/mol) | 60.10 |
| MDL Number | MFCD00002941 |
| InChI Key | BDERNNFJNOPAEC-UHFFFAOYSA-N |
| Synonym | 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol |
| PubChem CID | 1031 |
| ChEBI | CHEBI:28831 |
| IUPAC Name | propan-1-ol |
| SMILES | CCCO |
Chemical Identifiers
| 71-23-8 | |
| 60.10 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| 1031 | |
| propan-1-ol |
| C3H8O | |
| MFCD00002941 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CHEBI:28831 | |
| CCCO |
Specifications
| 71-23-8 | |
| 100.0000% | |
| 97.2°C | |
| 23°C | |
| C3H8O | |
| UN1274 | |
| 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol | |
| CCCO | |
| 60.10 | |
| CHEBI:28831 | |
| >99.5% | |
| n-Propyl Alcohol, B&J Brand™, for HPLC, GC and spectrophotometry, >99.5% |
| 0.5 AU max. at 225 nm, 0.05 AU max. at 250 nm, 0.005 AU max. at 300 nm, 0.005 AU max. at 400 nm | |
| 0.8053 g/cm3 | |
| 4 x 4 L | |
| Glass Bottle | |
| MFCD00002941 | |
| 1098242 | |
| BDERNNFJNOPAEC-UHFFFAOYSA-N | |
| propan-1-ol | |
| 1031 | |
| 60.09 | |
| HPLC |
Safety and Handling
ShelfLife : 547 days from date of manufacture
DOTInformation : Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: n-Propanol