missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl acetate, 99%

Catalog No. p-7046626
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL15355AP 500 mL
AAL15355AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL15355AP Supplier Thermo Scientific Chemicals Supplier No. L15355AP
Only null left

CAS: 109-60-4 | C5H10O2 | 102.13 g/mol

n-propyl acetate is used as a solvent. It plays an important role in the printing inks industry are flexographic and special screen printing inks. It is widely used in fragrances and as a flavor additive due to its odor. It acts as a good solvent for cellulose nitrate, acrylates, alkyd resins, rosin, plasticizers, waxes, oils and fats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-propyl acetate is used as a solvent. It plays an important role in the printing inks industry are flexographic and special screen printing inks. It is widely used in fragrances and as a flavor additive due to its odor. It acts as a good solvent for cellulose nitrate, acrylates, alkyd resins, rosin, plasticizers, waxes, oils and fats.

Solubility
Miscible with alcohols, ketones, aldehydes, ethers, glycols and glycol ethers. Slightly soluble in water.

Notes
Incompatible with oxidizing agents.

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
MDL Number MFCD00009372
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
Synonym n-propyl acetate, acetic acid, propyl ester, propyl ethanoate, 1-acetoxypropane, n-propyl ethanoate, 1-propyl acetate, acetic acid n-propyl ester, octan propylu, propylacetate, acetic acid propyl ester
PubChem CID 7997
ChEBI CHEBI:40116
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

Melting Point -92°C
Density 0.887
Boiling Point 99°C to 102°C
Flash Point 14°C (57°F)
Odor Fruit-like
Refractive Index 1.384
Linear Formula CH3CO2CH2CH2CH3
Quantity 500 mL
UN Number UN1276
Beilstein 1740764
Merck Index 14,7841
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,glycols and glycol ethers. Slightly soluble in water.
Formula Weight 102.13
Percent Purity 99%
Chemical Name or Material n-Propyl acetate
Show More Show Less
Hazard Category H225-H319-H336
Hazard Statement GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: N-PROPYL ACETATE
EINECSNumber 203-686-1
RTECSNumber AJ3675000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only