missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Acetate, 99%, Spectrum™ Chemical

Catalog No. 18612043
Change view
Click to view available options
Quantity:
500 mL
2.5 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly-Coated Glass Bottle
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612043 200 L Drum
18612044 500 mL Amber Glass Bottle
18612041 2.5 L Poly-Coated Glass Bottle
18612042 20 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612043 Supplier Spectrum Chemical Mfg Cor Supplier No. P1612200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 109-60-4
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.13
InChI Key YKYONYBAUNKHLG-UHFFFAOYSA-N
IUPAC Name propyl acetate
SMILES CCCOC(C)=O

Specifications

CAS Max % 1
Packaging Drum
Quantity 200 L
Percent Purity 0.99
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.