missing translation for 'onlineSavingsMsg'
Learn More

n-Propyl Acetate, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor P161220LT

 View more versions of this product

Catalog No. 18-612-042


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

109-60-4
Steel Can
20 L
CCCOC(C)=O
102.13
Reagent
1
C5H10O2
YKYONYBAUNKHLG-UHFFFAOYSA-N
propyl acetate
0.99

Chemical Identifiers

109-60-4
102.13
propyl acetate
C5H10O2
YKYONYBAUNKHLG-UHFFFAOYSA-N
CCCOC(C)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.