missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Propionylglycine, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 21709-90-0 |
| Linear Formula | C5H9 NO3 |
| MDL Number | MFCD09049674 |
| Synonym | 2-(Propionylamino)acetic acid; N-(1-Oxopropyl)glycine; NSC 158539; Propionyl glycine |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 131.13 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |