missing translation for 'onlineSavingsMsg'
Learn More

N-Propionylglycine, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013553
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013553 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013553 Supplier Merck Emd Millipore Supplier No. 9298610MG
Only null left

Specifications

CAS 21709-90-0
Linear Formula C5H9 NO3
MDL Number MFCD09049674
Synonym 2-(Propionylamino)acetic acid; N-(1-Oxopropyl)glycine; NSC 158539; Propionyl glycine
Type Analytical Standard
Quantity 10 mg
Formula Weight 131.13
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard