missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Propargylphthalimide 98.0+%, TCI America™
Supplier: TCI America P232925G
Specifications
| N-Propargylphthalimide | |
| 153°C | |
| MFCD00065028 | |
| n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl | |
| O=C1N(CC#C)C(=O)C2=CC=CC=C12 | |
| 185.18 | |
| 185.18 | |
| Crystalline Powder |
| 7223-50-9 | |
| C11H7NO2 | |
| 25 g | |
| PAZCLCHJOWLTGA-UHFFFAOYSA-N | |
| 2-(prop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione | |
| 81644 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 7223-50-9 | |
| 185.18 | |
| PAZCLCHJOWLTGA-UHFFFAOYSA-N | |
| 81644 | |
| O=C1N(CC#C)C(=O)C2=CC=CC=C12 |
| C11H7NO2 | |
| MFCD00065028 | |
| n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl | |
| 2-(prop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione |
Safety and Handling
TSCA : No