missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Propargylphthalimide 98.0+%, TCI America™
$316.89 - $993.54
Chemical Identifiers
| CAS | 7223-50-9 |
|---|---|
| Molecular Formula | C11H7NO2 |
| Molecular Weight (g/mol) | 185.18 |
| MDL Number | MFCD00065028 |
| InChI Key | PAZCLCHJOWLTGA-UHFFFAOYSA-N |
| Synonym | n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl |
| PubChem CID | 81644 |
| IUPAC Name | 2-(prop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione |
| SMILES | O=C1N(CC#C)C(=O)C2=CC=CC=C12 |
Chemical Identifiers
| 7223-50-9 | |
| 185.18 | |
| PAZCLCHJOWLTGA-UHFFFAOYSA-N | |
| 81644 | |
| O=C1N(CC#C)C(=O)C2=CC=CC=C12 |
| C11H7NO2 | |
| MFCD00065028 | |
| n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl | |
| 2-(prop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione |
Specifications
| 7223-50-9 | |
| C11H7NO2 | |
| 5 g | |
| PAZCLCHJOWLTGA-UHFFFAOYSA-N | |
| 2-(prop-2-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione | |
| 81644 | |
| ≥98.0% (GC) | |
| N-Propargylphthalimide |
| 153°C | |
| MFCD00065028 | |
| n-propargylphthalimide, 2-prop-2-yn-1-yl isoindoline-1,3-dione, n-prop-2-ynylphthalimide, 2-2-propynyl-1h-isoindole-1,3 2h-dione, 2-prop-2-ynyl-isoindole-1,3-dione, n-propargyl phthalimide, 1h-isoindole-1,3 2h-dione, 2-2-propynyl, 2-prop-2-ynyl isoindoline-1,3-dione, 2-prop-2-yn-1-yl isoindole-1,3-dione, 1h-isoindole-1,3 2h-dione,2-2-propyn-1-yl | |
| O=C1N(CC#C)C(=O)C2=CC=CC=C12 | |
| 185.18 | |
| 185.18 | |
| Crystalline Powder |
Safety and Handling
TSCA : No