missing translation for 'onlineSavingsMsg'
Learn More

N-Phthalyl-L-tryptophan 98.0+%, TCI America™

Catalog No. p-7135365
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P202350MG 50 mg
P2023200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P202350MG Supplier TCI America Supplier No. P202350MG
Only null left

Chemical Identifiers

CAS 48208-26-0
Molecular Formula C19H14N2O4
Molecular Weight (g/mol) 334.331
MDL Number MFCD08705332
InChI Key HPTXLHAHLXOAKV-MRXNPFEDSA-N
Synonym 2r-2-1,3-dioxoisoindol-2-yl-3-1h-indol-3-yl propanoic acid, r-2-1,3-dioxoisoindoline-2-yl-3-1h-indole-3-yl propanoic acid
PubChem CID 702560
IUPAC Name (2R)-2-(1,3-dioxoisoindol-2-yl)-3-(1H-indol-3-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)C(CC3=CNC4=CC=CC=C43)C(=O)O

Specifications

Melting Point 181°C
Color Yellow
Quantity 50 mg
Formula Weight 334.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phthalyl-L-tryptophan
TSCA No
Recommended Storage Freezer