Learn More
N-Phthaloylglycine, 98+%
CAS: 4702-13-0 | C10H7NO4 | 205.169 g/mol
$107.52 - $1150.97
Chemical Identifiers
| CAS | 4702-13-0 |
|---|---|
| Molecular Formula | C10H7NO4 |
| Molecular Weight (g/mol) | 205.169 |
| MDL Number | MFCD00005900 |
| InChI Key | WQINSVOOIJDOLJ-UHFFFAOYSA-N |
| Synonym | n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid |
| PubChem CID | 20825 |
| IUPAC Name | 2-(1,3-dioxoisoindol-2-yl)acetic acid |
| SMILES | C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1592018
|
Thermo Scientific Chemicals
A1592018 |
50 g |
Each for $107.52
|
|
|||||
|
AAA1592030
|
Thermo Scientific Chemicals
A1592030 |
250 g |
Each for $404.26
|
|
|||||
|
AAA159200B
|
Thermo Scientific Chemicals
A159200B |
1000 g |
Each for $1,150.97
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4702-13-0 | |
| 205.169 | |
| WQINSVOOIJDOLJ-UHFFFAOYSA-N | |
| 20825 | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O |
| C10H7NO4 | |
| MFCD00005900 | |
| n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid | |
| 2-(1,3-dioxoisoindol-2-yl)acetic acid |
Specifications
| 194°C to 198°C | |
| C10H7NO4 | |
| 184174 | |
| WQINSVOOIJDOLJ-UHFFFAOYSA-N | |
| 2-(1,3-dioxoisoindol-2-yl)acetic acid | |
| 205.169 | |
| 205.17 | |
| N-Phthaloylglycine |
| 4702-13-0 | |
| MFCD00005900 | |
| n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O | |
| 50 g | |
| 20825 | |
| ≥98% |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 225-177-3
RTECSNumber : NR3439000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only