Learn More
N-Phthaloylglycine, 98+%
CAS: 4702-13-0 | C10H7NO4 | 205.169 g/mol
Supplier: Thermo Scientific Chemicals A1592030
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 194°C to 198°C | |
| 4702-13-0 | |
| MFCD00005900 | |
| n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O | |
| 250 g | |
| 20825 | |
| ≥98% |
| N-Phthaloylglycine | |
| C10H7NO4 | |
| 184174 | |
| WQINSVOOIJDOLJ-UHFFFAOYSA-N | |
| 2-(1,3-dioxoisoindol-2-yl)acetic acid | |
| 205.169 | |
| 205.17 |
Chemical Identifiers
| 4702-13-0 | |
| 205.169 | |
| WQINSVOOIJDOLJ-UHFFFAOYSA-N | |
| 20825 | |
| C1=CC=C2C(=C1)C(=O)N(C2=O)CC(=O)O |
| C10H7NO4 | |
| MFCD00005900 | |
| n-phthaloylglycine, phthalimidoacetic acid, n-phthalylglycine, phthaloylglycine, n-carboxymethyl phthalimide, n,n-phthaloylglycine, n-phthalylglycin, 1,3-dioxo-2-isoindolineacetic acid, 1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl acetic acid, 2-1,3-dioxoisoindolin-2-yl acetic acid | |
| 2-(1,3-dioxoisoindol-2-yl)acetic acid |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H500
EINECSNumber : 225-177-3
RTECSNumber : NR3439000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only