missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phthaloyl-DL-glutamic Anhydride 98.0+%, TCI America™
Supplier: TCI America P178925G
Specifications
| N-Phthaloyl-DL-glutamic Anhydride | |
| 205°C | |
| C13H9NO5 | |
| 25 g | |
| ICDLEMPZXFCQEB-UHFFFAOYNA-N | |
| 2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione | |
| 92193 | |
| ≥98.0% (T) |
| 3343-28-0 | |
| White | |
| MFCD00005886 | |
| 2-Phthalimidoglutaric Anhydride | |
| O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12 | |
| 259.22 | |
| 259.22 | |
| Crystalline Powder |
Chemical Identifiers
| 3343-28-0 | |
| 259.22 | |
| ICDLEMPZXFCQEB-UHFFFAOYNA-N | |
| 92193 | |
| O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12 |
| C13H9NO5 | |
| MFCD00005886 | |
| 2-Phthalimidoglutaric Anhydride | |
| 2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione |
Safety and Handling
RTECSNumber : MA3900000
TSCA : Yes