missing translation for 'onlineSavingsMsg'
Learn More

N-Phthaloyl-DL-glutamic Anhydride 98.0+%, TCI America™

$216.24 - $825.78

Chemical Identifiers

CAS 3343-28-0
Molecular Formula C13H9NO5
Molecular Weight (g/mol) 259.22
MDL Number MFCD00005886
InChI Key ICDLEMPZXFCQEB-UHFFFAOYNA-N
Synonym 2-Phthalimidoglutaric Anhydride
PubChem CID 92193
IUPAC Name 2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
P17895G
View Documents
TCI America
P17895G
5 g N/A
Only null left
 
P178925G
View Documents
TCI America
P178925G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

3343-28-0
259.22
ICDLEMPZXFCQEB-UHFFFAOYNA-N
92193
O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12
C13H9NO5
MFCD00005886
2-Phthalimidoglutaric Anhydride
2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione

Specifications

3343-28-0
White
MFCD00005886
2-Phthalimidoglutaric Anhydride
O=C1N(C2CCC(=O)OC2=O)C(=O)C2=CC=CC=C12
259.22
259.22
Crystalline Powder
205°C
C13H9NO5
5 g
ICDLEMPZXFCQEB-UHFFFAOYNA-N
2-(2,6-dioxooxan-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
92193
≥98.0% (T)
N-Phthaloyl-DL-glutamic Anhydride
Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA3900000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'