missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylthiourea, 97%

Catalog No. AAL0669009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0669009 10 g
AAL0669018 50 g
AAL0669030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0669009 Supplier Thermo Scientific Chemicals Supplier No. L0669009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 103-85-5 | C7H8N2S | 152.22 g/mol

N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-phenylthiourea is used in paternity testing prior to the advent of DNA testing. It inhibits melanogenesis and is used to grow transparent fish. It is also used as a repellent for rat, rabbits, weasels and also in medical genetics.

Solubility
Soluble in hot water and alcohol.

Notes
Incompatible with strong oxidizing agents, strong acids and strong bases.

Chemical Identifiers

CAS 103-85-5
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.22
MDL Number MFCD00004933
InChI Key FULZLIGZKMKICU-UHFFFAOYSA-N
Synonym n-phenylthiourea, 1-phenylthiourea, 1-phenyl-2-thiourea, phenylthiocarbamide, thiourea, phenyl, phenyl-2-thiourea, fenylthiomocovina, urea, 1-phenyl-2-thio, rcra waste number p093, phenyl-thiourea
PubChem CID 676454
ChEBI CHEBI:46261
IUPAC Name phenylthiourea
SMILES NC(=S)NC1=CC=CC=C1

Specifications

Melting Point 149°C to 153°C
Odor Odorless
Quantity 10 g
UN Number UN2811
Beilstein 907309
Merck Index 14,7315
Solubility Information Soluble in hot water and alcohol.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material N-Phenylthiourea
Hazard Category H300-H317
Hazard Statement GHS H Statement
H300
Fatal if swallowed.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P310-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 203-151-2
RTECSNumber YU1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only