missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylthiobenzamide 98.0+%, TCI America™
$275.87 - $275.87
Chemical Identifiers
| CAS | 636-04-4 |
|---|---|
| Molecular Formula | C13H11NS |
| Molecular Weight (g/mol) | 213.298 |
| MDL Number | MFCD00022147 |
| InChI Key | BOQKCADLPNLYCZ-UHFFFAOYSA-N |
| Synonym | Thiobenzanilide |
| PubChem CID | 668263 |
| IUPAC Name | N-phenylbenzenecarbothioamide |
| SMILES | C1=CC=C(C=C1)C(=S)NC2=CC=CC=C2 |
Chemical Identifiers
| 636-04-4 | |
| 213.298 | |
| BOQKCADLPNLYCZ-UHFFFAOYSA-N | |
| 668263 | |
| C1=CC=C(C=C1)C(=S)NC2=CC=CC=C2 |
| C13H11NS | |
| MFCD00022147 | |
| Thiobenzanilide | |
| N-phenylbenzenecarbothioamide |
Specifications
| 636-04-4 | |
| Yellow | |
| MFCD00022147 | |
| Thiobenzanilide | |
| C1=CC=C(C=C1)C(=S)NC2=CC=CC=C2 | |
| 213.298 | |
| 213.30 | |
| Crystalline Powder |
| 102°C | |
| C13H11NS | |
| 5 g | |
| BOQKCADLPNLYCZ-UHFFFAOYSA-N | |
| N-phenylbenzenecarbothioamide | |
| 668263 | |
| ≥98.0% (HPLC,N) | |
| N-Phenylthiobenzamide |
Safety and Handling
RTECSNumber : CV8925000
TSCA : Yes