missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylsuccinimide 98.0+%, TCI America™
Supplier: TCI America P135925G
Specifications
| N-Phenylsuccinimide | |
| 155°C | |
| C10H9NO2 | |
| 25 g | |
| C1CC(=O)N(C1=O)C2=CC=CC=C2 | |
| 175.187 | |
| 175.19 | |
| Crystalline Powder |
| 83-25-0 | |
| White | |
| MFCD00022587 | |
| ZTUKZULGOCFJET-UHFFFAOYSA-N | |
| 1-phenylpyrrolidine-2,5-dione | |
| 66519 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 83-25-0 | |
| 175.187 | |
| ZTUKZULGOCFJET-UHFFFAOYSA-N | |
| 1-phenylpyrrolidine-2,5-dione |
| C10H9NO2 | |
| MFCD00022587 | |
| 66519 | |
| C1CC(=O)N(C1=O)C2=CC=CC=C2 |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator