accessibility menu, dialog, popup

UserName

CAS RN 83-25-0

OON

CAS RN 83-25-0

IUPAC Name: 1-phenylpyrrolidine-2,5-dione
Synonyms: N-phenylsuccinimide
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: ZTUKZULGOCFJET-UHFFFAOYSA-N
SMILES: O=C1CCC(=O)N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results