missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylsuccinimide 98.0+%, TCI America™

Catalog No. P13595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13595G 5 g
P135925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P13595G Supplier TCI America Supplier No. P13595G
Only null left

Chemical Identifiers

CAS 83-25-0
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.187
MDL Number MFCD00022587
InChI Key ZTUKZULGOCFJET-UHFFFAOYSA-N
PubChem CID 66519
IUPAC Name 1-phenylpyrrolidine-2,5-dione
SMILES C1CC(=O)N(C1=O)C2=CC=CC=C2

Specifications

Melting Point 155°C
Color White
Quantity 5 g
Formula Weight 175.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenylsuccinimide
TSCA No
Recommended Storage Refrigerator