missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyldiethanolamine 98.0+%, TCI America™

Catalog No. P0187500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0187500G 500 g
P018725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0187500G Supplier TCI America Supplier No. P0187500G

Chemical Identifiers

CAS 120-07-0
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00002845
InChI Key OJPDDQSCZGTACX-UHFFFAOYSA-N
Synonym n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
PubChem CID 8416
IUPAC Name 2-[(2-hydroxyethyl)(phenyl)amino]ethan-1-ol
SMILES OCCN(CCO)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Boiling Point 270°C
Quantity 500 g
Formula Weight 181.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Phenyldiethanolamine
EINECSNumber (3)-0160
RTECSNumber KM2100000
TSCA Yes