accessibility menu, dialog, popup

UserName

HONOH

CAS RN 120-07-0

IUPAC Name: 2-[(2-hydroxyethyl)(phenyl)amino]ethan-1-ol
Synonyms: phenyldiethanolamine
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: OJPDDQSCZGTACX-UHFFFAOYSA-N
SMILES: OCCN(CCO)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)