Learn More
N-Phenyldiethanolamine, 97%
CAS: 120-07-0 | C10H15NO2 | 181.24 g/mol
Supplier: Thermo Scientific Chemicals L139310E
Description
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.
Solubility
Soluble in water 3g/L @ 20ËC.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
Specifications
| N-Phenyldiethanolamine | |
| 55°C to 59°C | |
| 8.2 | |
| 178°C (352°F) | |
| C10H15NO2 | |
| MFCD00002845 | |
| 2096832 | |
| n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline | |
| OJPDDQSCZGTACX-UHFFFAOYSA-N | |
| 2-[N-(2-hydroxyethyl)anilino]ethanol | |
| 8416 | |
| 97% |
| 120-07-0 | |
| 1.1 | |
| 270°C | |
| Amine-like | |
| C6H5N(CH2CH2OH)2 | |
| 2500 g | |
| Air sensitive; Hygroscopic | |
| Soluble in water 3g/L @ 20?C. | |
| OCCN(CCO)C1=CC=CC=C1 | |
| 181.24 | |
| 181.23 |
Chemical Identifiers
| 120-07-0 | |
| 181.24 | |
| OJPDDQSCZGTACX-UHFFFAOYSA-N | |
| 8416 | |
| OCCN(CCO)C1=CC=CC=C1 |
| C10H15NO2 | |
| MFCD00002845 | |
| n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline | |
| 2-[N-(2-hydroxyethyl)anilino]ethanol |
Safety and Handling
GHS H Statement
H318-H373-H302
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P260-P264b-P270-P272-P280-P302+P352-P305+P351+P338-P307+P311-P310-P333+P313-P363-P501c
H317-H318-H370
EINECSNumber : 204-368-5
RTECSNumber : KM2100000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only