missing translation for 'onlineSavingsMsg'
Learn More

N-Phenyldiethanolamine, 97%

Catalog No. AAL1393122
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1393122 100 g
AAL1393136 500 g
AAL139310E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1393122 Supplier Thermo Scientific Chemicals Supplier No. L1393122
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 120-07-0 | C10H15NO2 | 181.24 g/mol

N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.

Solubility
Soluble in water 3g/L @ 20˚C.

Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.

Chemical Identifiers

CAS 120-07-0
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.24
MDL Number MFCD00002845
InChI Key OJPDDQSCZGTACX-UHFFFAOYSA-N
Synonym n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline
PubChem CID 8416
IUPAC Name 2-[N-(2-hydroxyethyl)anilino]ethanol
SMILES OCCN(CCO)C1=CC=CC=C1

Specifications

Melting Point 55°C to 59°C
Density 1.1
pH 8.2
Boiling Point 270°C
Flash Point 178°C (352°F)
Odor Amine-like
Linear Formula C6H5N(CH2CH2OH)2
Quantity 100 g
Beilstein 2096832
Sensitivity Air sensitive; Hygroscopic
Solubility Information Soluble in water 3g/L @ 20?C.
Formula Weight 181.23
Percent Purity 97%
Chemical Name or Material N-Phenyldiethanolamine
Show More Show Less
Hazard Category H317-H318-H370
Hazard Statement GHS H Statement
H318-H373-H302
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P272-P280-P302+P352-P305+P351+P338-P307+P311-P310-P333+P313-P363-P501c
EINECSNumber 204-368-5
RTECSNumber KM2100000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only