missing translation for 'onlineSavingsMsg'
Learn More

N-Phenylbenzohydroxamic acid, 98%

Catalog No. AC417370050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC417370050 5 g Glass bottle
AC417370250 25 g Glass bottle
AC417371000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC417370050 Supplier Thermo Scientific Chemicals Supplier No. 417370050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 304-88-1 | C13H11NO2 | 213.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 304-88-1
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.24
MDL Number MFCD00002111
InChI Key YLYIXDZITBMCIW-UHFFFAOYSA-N
Synonym n-benzoyl-n-phenylhydroxylamine, n-phenylbenzohydroxamic acid, n-hydroxybenzanilide, benzamide, n-hydroxy-n-phenyl, n-benzoyl-n-phenylhydroxamic acid, n-benzoylphenylhydroxylamine, benzohydroxamic acid, n-phenyl, tantalon, acmc-1cqm9, n-phenylbenzohydroxamicacid
PubChem CID 67536
IUPAC Name N-hydroxy-N-phenylbenzamide
SMILES ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 119°C to 123°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CON(OH)C6H5
Quantity 5 g
Formula Weight 213.24
Percent Purity 98%
Physical Form Needle-like Crystals or Crystalline Powder
Chemical Name or Material N-Phenylbenzohydroxamic acid
Show More Show Less
EINECSNumber 206-158-9

RUO – Research Use Only