accessibility menu, dialog, popup

UserName

OHNO

CAS RN 304-88-1

IUPAC Name: N-hydroxy-N-phenylbenzamide
Synonyms: N-hydroxy-N-phenylbenzamide
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: YLYIXDZITBMCIW-UHFFFAOYSA-N
SMILES: ON(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)