missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenyl-9-anthramine 98.0+%, TCI America™
$203.17 - $671.06
Chemical Identifiers
| CAS | 15424-38-1 |
|---|---|
| Molecular Formula | C20H15N |
| Molecular Weight (g/mol) | 269.35 |
| MDL Number | MFCD03093237 |
| InChI Key | VNXLMMFDPYSZKF-UHFFFAOYSA-N |
| Synonym | 9-Anilinoanthracene |
| PubChem CID | 13777361 |
| IUPAC Name | N-phenylanthracen-9-amine |
| SMILES | N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12 |
Chemical Identifiers
| 15424-38-1 | |
| 269.35 | |
| VNXLMMFDPYSZKF-UHFFFAOYSA-N | |
| 13777361 | |
| N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12 |
| C20H15N | |
| MFCD03093237 | |
| 9-Anilinoanthracene | |
| N-phenylanthracen-9-amine |
Specifications
| 15424-38-1 | |
| Yellow | |
| MFCD03093237 | |
| 9-Anilinoanthracene | |
| N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC2=CC=CC=C12 | |
| 269.35 | |
| 269.35 | |
| Crystalline Powder |
| 207°C | |
| C20H15N | |
| 1 g | |
| VNXLMMFDPYSZKF-UHFFFAOYSA-N | |
| N-phenylanthracen-9-amine | |
| 13777361 | |
| ≥98.0% (GC) | |
| N-Phenyl-9-anthramine |
Safety and Handling
TSCA : No