missing translation for 'onlineSavingsMsg'
Learn More

N-Phenacylpyridinium bromide, 97%, Thermo Scientific™

Catalog No. AC450120500
Change view
Click to view available options
Quantity:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC450120500 50g
AC450120100 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC450120500 Supplier Thermo Scientific Chemicals Supplier No. 450120500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16883-69-5
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.15
InChI Key PXSUMUYPXZEXDT-UHFFFAOYSA-M
Synonym 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
PubChem CID 2798954
IUPAC Name 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
SMILES C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Quantity 50g
Solubility Information Solubility in water: soluble.
Formula Weight 278.15
Percent Purity 97%
Chemical Name or Material N-Phenacylpyridinium bromide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
EINECSNumber 240-919-6

RUO – Research Use Only